(R)-4-Methyl-3-(hydroxymethyl)morpholine hydrochloride 95%

Lieferant: Apollo Scientific

OR55497-1G
APOSOR55497-1GEA 568 EUR
APOSOR55497-1G
(R)-4-Methyl-3-(hydroxymethyl)morpholine hydrochloride 95%
(R)-4-Methyl-3-(hydroxymethyl)morpholine hydrochloride
Formel: C₆H₁₃NO₂·ClH
Molecular Weight: 167,634 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26959188
CAS-Nummer: 1820569-49-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR