4-Methyl-2-(hydroxymethyl)morpholine 95%

Lieferant: Apollo Scientific

Synonyme: (4-Methyl-2-morpholinyl)methanol

OR55500-1G
APOSOR55500-1GEA 102 EUR
APOSOR55500-1G
4-Methyl-2-(hydroxymethyl)morpholine 95%
4-Methyl-2-(hydroxymethyl)morpholine
Formel: C₆H₁₃NO₂
Molecular Weight: 131,173 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12024541
CAS-Nummer: 40987-46-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR