(R)-4-Methyl-2-(hydroxymethyl)morpholine 95%

Lieferant: Apollo Scientific
Achtung

OR55502-1G
APOSOR55502-1GEA 192 EUR
APOSOR55502-1G
(R)-4-Methyl-2-(hydroxymethyl)morpholine 95%
(R)-4-Methyl-2-(hydroxymethyl)morpholine
Formel: C₆H₁₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD16294696
CAS-Nummer: 1159598-35-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR