1-Boc-6-methoxy-2-indolboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Methoxy-1H-indole-2-boronic acid, N-BOC protected, (1-(tert-Butoxycarbonyl)-6-methoxy-1H-indol-2-yl)boronic acid

OR5551-1G
APOSOR5551-1GEA 92.8 EUR
APOSOR5551-1G
1-Boc-6-methoxy-2-indolboronsäure 98%
1-Boc-6-methoxy-2-indolboronsäure
Formel: C₁₄H₁₈BNO₅
Molecular Weight: 291,11 g/mol
Schmelzpunkt: 100…103 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06659830
CAS-Nummer: 850568-65-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR