5-Brom-4-methoxy-2-nitroanilin 98%

Lieferant: Apollo Scientific

OR55519-1G OR55519-250MG
APOSOR55519-1GEA 232 EUR
APOSOR55519-1G APOSOR55519-250MG
5-Brom-4-methoxy-2-nitroanilin 98%
5-Brom-4-methoxy-2-nitroanilin
Formel: C₇H₇BrN₂O₃
Molecular Weight: 247.0461 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00031127
CAS-Nummer: 6943-69-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR