3-Bromo- N,N-dimethyl-4-methoxyaniline 95%

Lieferant: Apollo Scientific

OR55523-1G
APOSOR55523-1GEA 727 EUR
APOSOR55523-1G
3-Bromo- N,N-dimethyl-4-methoxyaniline 95%
3-Bromo- N,N-dimethyl-4-methoxyaniline
Formel: C₉H₁₂BrNO
Molecular Weight: 230,102 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16315291
CAS-Nummer: 102236-13-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR