2-Bromo-3-methoxy-6-nitroaniline 95%

Lieferant: Apollo Scientific

OR55533-1G
APOSOR55533-1GEA 1180 EUR
APOSOR55533-1G
2-Bromo-3-methoxy-6-nitroaniline 95%
2-Bromo-3-methoxy-6-nitroaniline
Formel: C₇H₇BrN₂O₃
Molecular Weight: 247,046 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22689544
CAS-Nummer: 90150-94-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR