4-Bromo-5-methoxy-1H-benzimidazole 95%
Lieferant: Apollo Scientific
OR55536-250MG
APOSOR55536-250MGEA
1090
EUR
APOSOR55536-250MG
4-Bromo-5-methoxy-1H-benzimidazole 95%
4-Bromo-5-methoxy-1H-benzimidazole
Formel:
C₈H₇BrN₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD22558065 CAS-Nummer: 1008361-66-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...