(R)-3-Cyclopropyl-2-hydroxypropanoic acid 95%

Lieferant: Apollo Scientific
Achtung

OR55546-250MG
APOSOR55546-250MGEA 1010 EUR
APOSOR55546-250MG
(R)-3-Cyclopropyl-2-hydroxypropanoic acid 95%
(R)-3-Cyclopropyl-2-hydroxypropanoic acid
Formel: C₆H₁₀O₃
Molecular Weight: 130,142 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17171014
CAS-Nummer: 174265-97-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR