2-Chloro-3-(trichloromethyl)pyridine 97%

Lieferant: Apollo Scientific
Achtung

OR55549-5G
APOSOR55549-5GEA 75.4 EUR
APOSOR55549-5G
2-Chloro-3-(trichloromethyl)pyridine 97%
2-Chloro-3-(trichloromethyl)pyridine
Formel: C₆H₃Cl₄N
Lagertemperatur: Raumtemperatur
MDL: MFCD00160150
CAS-Nummer: 72648-12-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR