3,5-Dibromo-4-chloroaniline 97%

Lieferant: Apollo Scientific
Achtung

OR55571-1G
APOSOR55571-1GEA 582 EUR
APOSOR55571-1G
3,5-Dibromo-4-chloroaniline 97%
3,5-Dibromo-4-chloroaniline
Formel: C₆H₄Br₂ClN
Molecular Weight: 285,364 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD18914595
CAS-Nummer: 35754-04-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR