2-Amino-4-(methoxycarbonyl)benzeneboronic acid pinacol ester hydrochloride 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 3-amino-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate hydrochloride

OR5560-1G
APOSOR5560-1GEA 292 EUR
APOSOR5560-1G
2-Amino-4-(methoxycarbonyl)benzeneboronic acid pinacol ester hydrochloride 95%
2-Amino-4-(methoxycarbonyl)benzeneboronic acid pinacol ester hydrochloride
Formel: C₁₄H₂₀BNO₄·HCl
Molecular Weight: 313,59 g/mol
Schmelzpunkt: 92…94 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06659918
CAS-Nummer: 850567-49-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR