4-(Benzyloxycarbonyl)-2-nitrobenzeneboronic acid 95%
Lieferant: Apollo Scientific
OR5561-1G
APOSOR5561-1GEA
268
EUR
APOSOR5561-1G
4-(Benzyloxycarbonyl)-2-nitrobenzeneboronic acid 95%
4-(Benzyloxycarbonyl)-2-nitrobenzeneboronic acid
Formel:
C₁₄H₁₂BNO₆ Molecular Weight: 301,059 g/mol Schmelzpunkt: 192…200 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02179461 CAS-Nummer: 850568-58-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...