4-(Bromomethyl)aniline, N-BOC protected 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
tert-Butyl (4-(bromomethyl)phenyl)carbamate
OR55614-1G
APOSOR55614-1GEA
144
EUR
APOSOR55614-1G
4-(Bromomethyl)aniline, N-BOC protected 97%
4-(Bromomethyl)aniline, N-BOC protected
Formel:
C₁₂H₁₆BrNO₂ Molecular Weight: 286,165 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11616718 CAS-Nummer: 239074-27-2 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...