3-(Oxan-4-yl)azetidine hydrochloride 95%

Lieferant: Apollo Scientific
Achtung

OR55621-100MG OR55621-250MG
APOSOR55621-100MGEA 0 EUR
APOSOR55621-100MG APOSOR55621-250MG
3-(Oxan-4-yl)azetidine hydrochloride 95%
3-(Oxan-4-yl)azetidine hydrochloride
Formel: C₈H₁₅NO·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD20483682
CAS-Nummer: 251569-98-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR