4-(1H-Imidazol-2-yl)benzylamine hydrochloride 99%

Lieferant: Apollo Scientific
Gefahr

OR55624-1G
APOSOR55624-1GEA 1900 EUR
APOSOR55624-1G
4-(1H-Imidazol-2-yl)benzylamine hydrochloride 99%
4-(1H-Imidazol-2-yl)benzylamine hydrochloride
Formel: C₁₀H₁₁N₃·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD28400887
UN: 3259
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR