Sodium-4-chlorobenzenesulphonate 98%

Lieferant: Apollo Scientific
Gefahr

OR55632-25G OR55632-500G
APOSOR55632-25GEA 50.9 EUR
APOSOR55632-25G APOSOR55632-500G
Sodium-4-chlorobenzenesulphonate 98%
Sodium-4-chlorobenzenesulphonate
Formel: C₆H₅ClO₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD00014731
CAS-Nummer: 5138-90-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR