1,5-Dimethyl-4-bromopyrazole-3-carbonitrile

Lieferant: Apollo Scientific
Gefahr

OR55637-1G OR55637-250MG
APOSOR55637-1GEA 574 EUR
APOSOR55637-1G APOSOR55637-250MG
1,5-Dimethyl-4-bromopyrazole-3-carbonitrile
1,5-Dimethyl-4-bromopyrazole-3-carbonitrile
Formel: C₆H₆BrN₃
Molecular Weight: 200.036 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30478437
CAS-Nummer: 863751-81-9
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR