6-Benzyloxy-1-boc-indole-2-boronic acid 98%
Lieferant: Apollo Scientific
Synonyme:
1H-Indole-1-carboxylicacid, 2-borono-6-(phenylmethoxy)-, 1,1-dimethylethyl ester, 6-Benzyloxy-1H-indole-2-boronic acid, N-BOC protected, 1-(TERT-BUTOXYCARBONYL)-6-(BENZYLOXY)-1H-INDOL-2-YL-2-BORONIC ACID
OR5564-1G
APOSOR5564-1GEA
248
EUR
APOSOR5564-1G
6-Benzyloxy-1-boc-indole-2-boronic acid 98%
6-Benzyloxy-1-boc-indole-2-boronic acid
Formel:
C₂₀H₂₂BNO₅ Molecular Weight: 367,208 g/mol Schmelzpunkt: 105…106 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06659831 CAS-Nummer: 850568-66-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...