N,1-Di-Boc-3-(4-bromophenyl)-3-azetidinamine 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR55649-1G
APOSOR55649-1GEA
1680
EUR
APOSOR55649-1G
N,1-Di-Boc-3-(4-bromophenyl)-3-azetidinamine 95%
N,1-Di-Boc-3-(4-bromophenyl)-3-azetidinamine
Formel:
C₁₉H₂₇BrN₂O₄ Molecular Weight: 427,333 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD31657914 CAS-Nummer: 1864803-36-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...