2-Chlor-3-nitrophenol 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR55653-1G
OR55653-25G
OR55653-5G
APOSOR55653-1GEA
21.9
EUR
APOSOR55653-1G
APOSOR55653-25G
APOSOR55653-5G
2-Chlor-3-nitrophenol 97%
2-Chlor-3-nitrophenol
Formel:
C₆H₄ClNO₃ Molecular Weight: 173.5500 g/mol Schmelzpunkt: 120 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11035882 CAS-Nummer: 603-84-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...