2-Chlor-3-nitrophenol 97%

Lieferant: Apollo Scientific
Achtung

OR55653-1G OR55653-25G OR55653-5G
APOSOR55653-1GEA 21.9 EUR
APOSOR55653-1G APOSOR55653-25G APOSOR55653-5G
2-Chlor-3-nitrophenol 97%
2-Chlor-3-nitrophenol
Formel: C₆H₄ClNO₃
Molecular Weight: 173.5500 g/mol
Schmelzpunkt: 120 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11035882
CAS-Nummer: 603-84-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR