6-Bromobenzofuran-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR55679-1G OR55679-250MG
APOSOR55679-1GEA 192 EUR
APOSOR55679-1G APOSOR55679-250MG
6-Bromobenzofuran-2-carboxylic acid
6-Bromobenzofuran-2-carboxylic acid
Formel: C₉H₅BrO₃
Molecular Weight: 241.038 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03012913
CAS-Nummer: 439107-94-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR