Ethyl-1-naphthoat 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR55681-50G
APOSOR55681-50GEA
253
EUR
APOSOR55681-50G
Ethyl-1-naphthoat 97%
Ethyl-1-naphthoat
Formel:
C₁₃H₁₂O₂ Molecular Weight: 200,24 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016805 CAS-Nummer: 3007-97-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...