Methyl mercaptan sodium salt-Lösung

Lieferant: Apollo Scientific
Gefahr

OR55697-1L OR55697-100G
APOSOR55697-1LEA 0 EUR
APOSOR55697-1L APOSOR55697-100G
Methyl mercaptan sodium salt-Lösung
Methyl mercaptan sodium salt
Formel: CH₃NaS
Lagertemperatur: Raumtemperatur
MDL: MFCD00174316
CAS-Nummer: 5188-07-8
UN: 2920
ADR: 8+3,I

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR