Cellopentaose

Lieferant: Apollo Scientific

Synonyme: D-(+)-Cellopentaose, Glc beta(1-4)Glc beta(1-4)Glc beta(1-4)Glc beta(1-4)Glc

OR5570T-25MG
APOSOR5570T-25MGEA 522 EUR
APOSOR5570T-25MG
Cellopentaose
Cellopentaose
Formel: C₃₀H₅₂O₂₆
Molecular Weight: 828,72 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00151164
CAS-Nummer: 2240-27-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR