Methyl-6-aminonicotinat 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 6-aminonicotinate, 6-Aminonicotinic acid methyl ester

OR5571-1G OR5571-25G OR5571-5G
APOSOR5571-1GEA 21.9 EUR
APOSOR5571-1G APOSOR5571-25G APOSOR5571-5G
Methyl-6-aminonicotinat 97%
Methyl-6-aminonicotinat
Formel: C₇H₈N₂O₂
Molecular Weight: 152.15 g/mol
Schmelzpunkt: 158…162 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00797844
CAS-Nummer: 36052-24-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR