(Chlormethyl)(isopropoxy)dimethylsilan 90%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Chloromethyl-dimethyl-propan-2-yloxysilane
OR55721-1ML
APOSOR55721-1MLEA
21.9
EUR
APOSOR55721-1ML
(Chlormethyl)(isopropoxy)dimethylsilan 90%
(Chloromethyl)isopropoxydimethylsilane
Formel:
C₆H₁₅ClOSi Molecular Weight: 166,72 g/mol Schmelzpunkt: 145 °C Flammpunkt: 28 °C |
MDL:
MFCD00010605 CAS-Nummer: 18171-11-4 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...