3-Bromo-2,6-dichlorophenol 97%

Lieferant: Apollo Scientific
Achtung

OR55726-1G
APOSOR55726-1GEA 417 EUR
APOSOR55726-1G
3-Bromo-2,6-dichlorophenol 97%
3-Bromo-2,6-dichlorophenol
Formel: C₆H₃BrCl₂O
Molecular Weight: 241.897 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17011993
CAS-Nummer: 116668-74-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR