3-Iodo-4-methylbenzyl alcohol
Lieferant: Apollo Scientific
|
Achtung
|
OR55730-1G
APOSOR55730-1GEA
0
EUR
APOSOR55730-1G
3-Iodo-4-methylbenzyl alcohol
3-Iodo-4-methylbenzyl alcohol
Formel:
C₈H₉IO Lagertemperatur: Raumtemperatur |
MDL:
MFCD02683557 CAS-Nummer: 165803-89-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...