2-Amino-3-brom-5-nitropyridin 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Brom-5-nitro-2-pyridylamin, 3-Brom-5-nitropyridin-2-amin

OR5574-5G
APOSOR5574-5GEA 0 EUR
APOSOR5574-5G
2-Amino-3-brom-5-nitropyridin 98%
2-Amino-3-brom-5-nitropyridin
Formel: C₅H₄BrN₃O₂
Molecular Weight: 218.01 g/mol
Schmelzpunkt: 218…222 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00955628
CAS-Nummer: 15862-31-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR