Pemetrexed disodium heptahydrate 97%

Lieferant: Apollo Scientific

Synonyme: Sodium (S)-2-(4-(2-(2-amino-4-oxo-4,7-dihydro-1H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl)benzamido)pentanedioate heptahydrate

OR55746-5G
APOSOR55746-5GEA 119 EUR
APOSOR55746-5G
Pemetrexed disodium heptahydrate 97%
Pemetrexed disodium heptahydrate
Formel: C₂₀H₁₉N₅O₆·2Na·7H₂O
Molecular Weight: 597.481 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09029394
CAS-Nummer: 357166-29-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR