1-(2-Pyrimidinyl)-3-pyrrolidinone

Lieferant: Apollo Scientific

OR55747-500MG
APOSOR55747-500MGEA 545 EUR
APOSOR55747-500MG
1-(2-Pyrimidinyl)-3-pyrrolidinone
1-(2-Pyrimidinyl)-3-pyrrolidinone
Formel: C₈H₉N₃O
Molecular Weight: 163,177 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20485714
CAS-Nummer: 956723-06-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR