3-Brom-2-chlor-5-nitropyridin 98%

Lieferant: Apollo Scientific
Gefahr

OR5575-100G
APOSOR5575-100GEA 161 EUR
APOSOR5575-100G
3-Brom-2-chlor-5-nitropyridin 98%
3-Bromo-2-chloro-5-nitropyridine
Formel: C₅H₂BrClN₂O₂
Molecular Weight: 237.44 g/mol
Schmelzpunkt: 67…68 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00233989
CAS-Nummer: 5470-17-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR