2-Formyl-5-methoxyphenylboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Methoxy-2-formylphenylboronic acid
OR5581-1G
OR5581-5G
OR5581-100MG
APOSOR5581-1GEA
125
EUR
APOSOR5581-1G
APOSOR5581-5G
APOSOR5581-100MG
2-Formyl-5-methoxyphenylboronsäure
2-Formyl-5-methoxyphenylboronsäure
Formel:
C₈H₉BO₄ Molecular Weight: 179.9657 g/mol Schmelzpunkt: 155…160 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03001335 CAS-Nummer: 40138-18-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...