5-Thiazolcarbonsäure 97%

Lieferant: Apollo Scientific
Achtung

OR5586-5G OR5586-1G OR5586-25G
APOSOR5586-5GEA 75.4 EUR
APOSOR5586-5G APOSOR5586-1G APOSOR5586-25G
5-Thiazolcarbonsäure 97%
5-Thiazolecarboxylic acid
Formel: C₄H₃NO₂S
Molecular Weight: 129.1371 g/mol
Schmelzpunkt: 190 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03428539
CAS-Nummer: 14527-41-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR