Cellotriose

Lieferant: Apollo Scientific
Achtung

Synonyme: (2R,3R,4R,5R)-4-(((2S,3R,4R,5S,6R)-3,4-Dihydroxy-6-(hydroxymethyl)-5-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-2,3,5,6-tetrahydroxyhexanal, Glc beta(1-4)Glc beta(1-4)Glc, D-(+)-Cellotriose

OR5590T-10MG
APOSOR5590T-10MGEA 210 EUR
APOSOR5590T-10MG
Cellotriose
Cellotriose
Formel: C₁₈H₃₂O₁₆
Molecular Weight: 504,44 g/mol
Schmelzpunkt: >165 °C (Zers.)
Lagertemperatur: Gefrierschrank
MDL: MFCD00069842
CAS-Nummer: 33404-34-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR