2-Acetamido-2-deoxy-D-glucono-1,5-lactone

Lieferant: Apollo Scientific

Synonyme: 2-Acetamido-2-deoxy-D-gluconolactone, N-[(3R,4R,5S,6R)-4,5-Dihydroxy-6-(hydroxymethyl)-2-oxotetrahydro-2H-pyran-3-yl]acetamide, N-Acetyl-D-glucosamino-1,5-lactone

OR5600T-25MG
APOSOR5600T-25MGEA 277 EUR
APOSOR5600T-25MG
2-Acetamido-2-deoxy-D-glucono-1,5-lactone
2-Acetamido-2-deoxy-D-glucono-1,5-lactone

Inhibits breakdown of products by b-hexosaminidase (s) when the culture medium is used as an enzyme source.

Formel: C₈H₁₃NO₆
Molecular Weight: 219,192 g/mol
Schmelzpunkt: 168…170 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00076154
CAS-Nummer: 19026-22-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR