2,5-Diiod-1-methylimidazol 97%

Lieferant: Apollo Scientific

Synonyme: 2,5-Diiod-1-methyl-1H-imidazol

OR5601-1G
APOSOR5601-1GEA 343 EUR
APOSOR5601-1G
2,5-Diiod-1-methylimidazol 97%
2,5-Diiodo-1-methylimidazole
Formel: C₄H₄I₂N₂
Molecular Weight: 333,9 g/mol
Schmelzpunkt: 138…143 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02179540
CAS-Nummer: 86026-81-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR