Methylthiazol-4-carboxylat 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 4-thiazolecarboxylate

OR5602-1G
APOSOR5602-1GEA 21.9 EUR
APOSOR5602-1G
Methylthiazol-4-carboxylat 97%
Methylthiazol-4-carboxylat
Formel: C₅H₅NO₂S
Molecular Weight: 143,1600 g/mol
Schmelzpunkt: 38…42 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06410940
CAS-Nummer: 59418-09-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR