Imidazol-4-carbonsäureethylester 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl imidazole-4-carboxylate

OR5606-1G OR5606-5G OR5606-25G
APOSOR5606-1GEA 85.8 EUR
APOSOR5606-1G APOSOR5606-5G APOSOR5606-25G
Imidazol-4-carbonsäureethylester 98%
Imidazol-4-carbonsäureethylester
Formel: C₆H₈N₂O₂
Schmelzpunkt: 160…162 °C
Lagertemperatur: Kühlschrank
MDL: MFCD07363812
CAS-Nummer: 23785-21-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR