Ethylthiazol-5-carboxylat 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl 5-thiazolecarboxylate
OR5607-5G
APOSOR5607-5GEA
55.1
EUR
APOSOR5607-5G
Ethylthiazol-5-carboxylat 98%
Ethylthiazol-5-carboxylat
Formel:
C₆H₇NO₂S Molecular Weight: 157.1900 g/mol Siedepunkt: 77…78 °C/2mmHg Schmelzpunkt: 14 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD06205069 CAS-Nummer: 32955-22-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...