Ethyl-2-[(tert-butoxycarbonyl)amino]-1,3-thiazole-5-carboxylate 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl 2-amino-1,3-thiazole-5-carboxylate, 2-BOC protected
OR5620-1G
APOSOR5620-1GEA
43.5
EUR
APOSOR5620-1G
Ethyl-2-[(tert-butoxycarbonyl)amino]-1,3-thiazole-5-carboxylate 98%
Ethyl-2-[(tert-butoxycarbonyl)amino]-1,3-thiazole-5-carboxylate
Formel:
C₁₁H₁₆N₂O₄S Molecular Weight: 272,33 g/mol Schmelzpunkt: 198…200 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD06409216 CAS-Nummer: 302964-01-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...