2,5-Dibrom-1-tritylimidazol 95%

Lieferant: Apollo Scientific

OR5621-1G
APOSOR5621-1GEA 303 EUR
APOSOR5621-1G
2,5-Dibrom-1-tritylimidazol 95%
2,5-Dibrom-1-tritylimidazol
Formel: C₂₂H₁₆Br₂N₂
Schmelzpunkt: 154…158 °C
Lagertemperatur: Kühlschrank
MDL: MFCD04115731
CAS-Nummer: 850429-53-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR