5-(Chloracetyl)oxindol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-(Chloroacetyl)-1,3-dihydro-2H-indol-2-one, 5-(2-Chloroacetyl)indolin-2-one

OR5624-1G
APOSOR5624-1GEA 21.9 EUR
APOSOR5624-1G
5-(Chloracetyl)oxindol 98%
5-(Chloracetyl)oxindol
Formel: C₁₀H₈ClNO₂
Molecular Weight: 209,6300 g/mol
Schmelzpunkt: 228…232 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04115734
CAS-Nummer: 65435-04-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR