5-(Chloracetyl)-6-chloroxindol 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
6-Chloro-5-(chloroacetyl)-1,3-dihydro-2H-indol-2-one
OR5625-1G
OR5625-25G
OR5625-5G
APOSOR5625-1GEA
48.2
EUR
APOSOR5625-1G
APOSOR5625-25G
APOSOR5625-5G
5-(Chloracetyl)-6-chloroxindol 97%
5-(Chloracetyl)-6-chloroxindol
Formel:
C₁₀H₇Cl₂NO₂ Schmelzpunkt: 211…213 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD04115735 CAS-Nummer: 118307-04-3 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...