1-Brom-4-(methylsulfonyl)-2-nitrobenzol 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR5627-1G
OR5627-5G
APOSOR5627-1GEA
123
EUR
APOSOR5627-1G
APOSOR5627-5G
1-Brom-4-(methylsulfonyl)-2-nitrobenzol 98%
1-Bromo-4-(methylsulphonyl)-2-nitrobenzene
Formel:
C₇H₆BrNO₄S Schmelzpunkt: 114…117 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00054605 CAS-Nummer: 94832-06-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...