4-(Cyanmethyl)tetrahydropyran 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR5629-1G
APOSOR5629-1GEA
54.4
EUR
APOSOR5629-1G
4-(Cyanmethyl)tetrahydropyran 95%
4-(Cyanmethyl)tetrahydropyran
Formel:
C₇H₁₁NO Siedepunkt: 83…85 °C/0,5mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD04973733 CAS-Nummer: 850429-50-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...