Cyclohexylhydrazine hydrochloride

Lieferant: Apollo Scientific
Gefahr

OR5641-1G OR5641-25G
APOSOR5641-1GEA 0 EUR
APOSOR5641-1G APOSOR5641-25G
Cyclohexylhydrazine hydrochloride
Cyclohexylhydrazine hydrochloride
Formel: C₆H₁₄N₂
Schmelzpunkt: 110…114 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00060160
CAS-Nummer: 24214-73-1
UN: 3263
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR