Pyrrolidin-1-ylacetic acid hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(Pyrrolidin-1-yl)acetic acid hydrochloride
OR5655-1G
APOSOR5655-1GEA
23.2
EUR
APOSOR5655-1G
Pyrrolidin-1-ylacetic acid hydrochloride
Pyrrolidin-1-ylacetic acid hydrochloride
Formel:
C₆H₁₂ClNO₂ Molecular Weight: 165,62 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06801274 CAS-Nummer: 6628-74-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...